Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM106A Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501732904 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2904 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2904 Supplier Proteintech Group Inc Supplier No. 204601AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Transmembrane protein 106A (TMEM106A) is a single-pass transmembrane protein that is closely related to TMEM106B, a protein that is thought to be a novel risk factor for frontotemporal lobar degeneration (FTLD), a group of clinically, pathologically and genetically heterogeneous disorders associated with atrophy in the frontal lobe and temporal lobe of the brain. The actual roles of TMEM106A and TMEM106B are still undetermined; however, as TMEM106B is involved in FTLD, it is possible that TMEM106A may also be a risk factor for FTLD.

Specifications

Antigen TMEM106A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TMEM106A
Gene Accession No. Q96A25
Gene Alias TMEM106A, transmembrane protein 106A
Gene Symbols TMEM106A
Host Species Rabbit
Immunogen TMEM106A Fusion Protein Ag14294
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113277
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.