Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM119 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735707 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5707 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5707 Supplier Proteintech Group Inc Supplier No. 275851AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

27585-1-AP recognizes a 45-50 kDa band likely due to glycosylation.

TMEM119 immunohistochemistry might provide a useful tool for investigating the biology and pathology of human microglia. The predicted MW of TMEM119 is 29 kDa.

Specifications

Antigen TMEM119
Applications Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TMEM119
Gene Accession No. Q4V9L6
Gene Alias OBIF, Osteoblast induction factor, TMEM119, transmembrane protein 119, UNQ731/PRO1415
Gene Symbols TMEM119
Host Species Rabbit
Immunogen TMEM119 Fusion Protein Ag26269
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338773
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.