Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tmem125 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110807 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110807 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110807 Supplier Invitrogen™ Supplier No. PA5110807
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSEQEAQAPG GRGLPPDMLA EQVELWWSQQ P Highest antigen sequence indentity to the following orthologs: Rat - 71%, Mouse - 71%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Tmem125
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tmem125
Gene Accession No. Q96AQ2
Gene Alias 6330530A05Rik; AV223516; TMEM125; Transmembrane protein 125
Gene Symbols Tmem125
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Tmem125. Recombinant protein control fragment (Product #RP-91185).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128218
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.