Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM135 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564416
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564416 Supplier Invitrogen™ Supplier No. PA564416
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SALRFIVGKE EIPTHSFSPE AAYAKVEQKR EQHEEKPRRM NMIGLVRKFV DSICKHGPRH RCCKHYEDNC Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

The transmembrane protein 135 (TMEM135), also known as peroxisomal membrane protein 52 (PMP52), was initially identified as a protein that might be critical for adipogenesis and osteoblastogenesis. Further studies have indicated that TMEM135 may be part of a regulatory circuit that plays an important role in fat metabolism and energy expenditure in both C. elegans as well as mammalian organisms. Finally, recent experiments has suggested that TMEM135 may be an additional driver of breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM135
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM135
Gene Accession No. Q86UB9
Gene Alias 2810439K08Rik; AW319712; Peroxisomal membrane protein 52; PMP52; RGD1309948; TMEM135; Transmembrane protein 135
Gene Symbols TMEM135
Host Species Rabbit
Immunogen Recombinant protein fragment of Human TMEM135 (Product #RP-103913)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65084
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.