Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM150B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571527
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571527 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571527 Supplier Invitrogen™ Supplier No. PA571527
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM150B Polyclonal Antibody for Western Blot, IHC (P), ICC/IF

Modulator of macroautophagy that causes accumulation of autophagosomes under basal conditions and enhances autophagic flux. Represses cell death and promotes long-term clonogenic survival of cells grown in the absence of glucose in a macroautophagy-independent manner. May have some role in extracellular matrix engulfment or growth factor receptor recycling, both of which can modulate cell survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM150B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TMEM150B
Gene Accession No. A6NC51
Gene Alias hCG1651476; Modulator of macroautophagy TMEM150B; Protein DRAM-3; TMEM150B; TMEM224; Transmembrane protein 150B; transmembrane protein 224; transmembrane protein LOC284417
Gene Symbols TMEM150B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 13-41 amino acids from the N-terminal region of human TMEM150B.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284417
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.