Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM163 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114329
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114329 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114329 Supplier Invitrogen™ Supplier No. PA5114329
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM163 Polyclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot

May bind zinc and other divalent cations and recruit them to vesicular organelles. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM163
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene TMEM163
Gene Accession No. A9CMA6, Q8C996, Q8TC26
Gene Alias 2610024A01Rik; DC29; RGD1306212; Sv31; synaptic vesicle membrane protein of 31 kDa; synaptic vesicle protein of 30 kD; Tmem163; Transmembrane protein 163
Gene Symbols TMEM163
Host Species Rabbit
Immunogen Synthetic peptide within Human TMEM163 aa 55-104.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360839, 72160, 81615
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.