Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM176A Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7209470 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2856 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2856 Supplier Proteintech Group Inc Supplier No. 203781AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function of this protein remains unknown.

Specifications

Antigen TMEM176A
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TMEM176A
Gene Accession No. Q96HP8
Gene Alias GS188, HCA112, TMEM176A, transmembrane protein 176A
Gene Symbols TMEM176A
Host Species Rabbit
Immunogen TMEM176A Fusion Protein Ag14194
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55365
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.