Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM230 Recombinant Rabbit Monoclonal Antibody (24H23L8)

Catalog No. 702792
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-279-2 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-279-2 Supplier Invitrogen™ Supplier No. 702792
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Rat, Pig. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Involved in trafficking and recycling of synaptic vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM230
Applications Western Blot
Classification Recombinant Monoclonal
Clone 24H23L8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene TMEM230
Gene Accession No. Q96A57
Gene Alias 5730494N06Rik; AA407821; C20orf30; dJ1116H23.2.1; HSPC274; HSPC274 protein; RGD1307399; TMEM230; Transmembrane protein 230; UNQ2432/PRO4992; UPF0414 transmembrane protein C20orf30; UPF0414 transmembrane protein C20orf30 homolog
Gene Symbols TMEM230
Host Species Rabbit
Immunogen Peptides corresponding to human TMEM230 [1) aa101-aa120 2) aa26-aa45].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29058
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.