Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM48 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116770
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116770 Supplier Invitrogen™ Supplier No. PA5116770
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: MCF7, Jurkat, HeLa, Mouse heart, Mouse brain Immunogen sequence: QYSPSRRQEV FSLSQPGGHP HNWTAISREC LNLLNGMTQK LILYQEAAAT NGRVSSSYPV EPKKLNSPEE TAFQTPKSSQ MPRPSVPPLV KTSLFSSKLS TPDVVSPFGT PFGSSVMNRM AGIFDVNTCY GSPQSPQLIR RGPRLWTSAS DQQMTEFSNP SPSTSISAEG KTMRQPSVIY SWIQNKREQI K.

TMEM48 is a component of the nuclear pore complex (NPC), which plays a key role in de novo assembly and insertion of NPC in the nuclear envelope. TMEM48 is required for NPC and nuclear envelope assembly, possibly by forming a link between the nuclear envelope membrane and soluble nucleoporins, thereby anchoring the NPC in the membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM48
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene NDC1
Gene Accession No. Q6AXN4, Q8VCB1, Q9BTX1
Gene Alias 2810475A17Rik; AI450313; FLJ10407; FLJ12556; FLJ34120; hNDC1; ndc1; NDC1 transmembrane nucleoporin; NET3; nuclear division cycle 1 homolog; nucleoporin NDC1; RP4-654H19.1; tmem48; Transmembrane protein 48; wu:fk93f04; zgc:55636
Gene Symbols NDC1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 340-530 of human NDC1 (NP_0605573).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362557, 55706, 72787
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.