Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM57 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585898
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585898 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585898 Supplier Invitrogen™ Supplier No. PA585898
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM57 Polyclonal Antibody for Western Blot

Plays a role in the regulation of neuronal activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM57
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene TMEM57
Gene Accession No. Q7TQE6, Q8N5G2
Gene Alias 1110007C24Rik; 9230118A01Rik; AI317300; AI606104; brain-specific adapter protein C61; brain-specific adaptor protein C61; C61; MACO1; macoi; Macoilin; Macoilin-1; Tmem57; Transmembrane protein 57
Gene Symbols TMEM57
Host Species Rabbit
Immunogen Synthetic peptide within Human TMEM57 aa 122-171.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55219, 66146
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.