Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TMEM59 Recombinant Protein Antigen
SDP

Catalog No. NB171670 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171670 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171670 Supplier R&D Systems™ Supplier No. NBP317950PEP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Recombinant Protein

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 9528
Molecular Weight (g/mol) M.W. Theoretical: 29 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias C1orf8, chromosome 1 open reading frame 8, FLJ42977, HSPC001, Liver membrane-bound protein, transmembrane protein 59
Gene Symbol TMEM59
Research Category Alzheimers Research, Neurodegeneration, Neuroscience, Signal Transduction
Product Line Novus
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein TMEM59
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.