Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM86A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563403 Supplier Invitrogen™ Supplier No. PA563403
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VALSAVESRE PVEHYRLTKA N Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 81%.

TMEM86A gene ontology annotations related to this gene include ether lipid metabolic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM86A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM86A
Gene Accession No. Q8N2M4
Gene Alias 1810054O13Rik; AI414959; AW742698; hypothetical protein LOC532018; im:7137034; Lysoplasmalogenase-like protein TMEM86A; RGD1305749; tmem86a; Transmembrane protein 86A; zgc:113571
Gene Symbols TMEM86A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM86A. Recombinant protein control fragment (Product #RP-101820).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144110
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.