Learn More
Enzo Life Sciences TMP-153 (20mg). CAS: 128831-46-9
SDP
Supplier:  Enzo Life Sciences BMLEI3170020
Encompass_Preferred
TMP-153 is a potent and selective inhibitor of acyl-CoA:cholesterol acyltransferase (ACAT), IC50=5-10 nM. It inhibits cholesterol absorption and lowers plasma cholesterol in rats and hamsters. ACAT inhibitors improve plasma lipid profile and ameliorate proteinuria in nephrotic animals. Purity: ≥98%. Solubility: Soluble in DMSO (>25mg/ml) or ethanol (10mg/ml with warming). Long Term Storage: Ambient.
Catalog No. 50-201-0705
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.