Learn More
Medchemexpress LLC HY-102075A 5mg Medchemexpress, TMP778 CAS:1422053-04-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY102075A5MG
Encompass_Preferred
Medchemexpress, HY-102075A 5mg TMP778 CAS:1422053-04-0 TMP778 is a potent and selevtive RORγt inverse agonist, with an IC50 of 7 nM in FRET assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0649
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.