Learn More
Santa Cruz Biotechnology Anti-TMPRSS2 Antibody (H-4), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC515727
Encompass
Anti-TMPRSS2 Antibody (H-4) is recommended for detection of TMPRSS2 catalytic chain of mouse, rat and human origin by WB, IP, IF, IHC(P) and ELISA.
Catalog No. NC1946350
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.