Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TMSI Silylation Reagent

Catalog No. PI88623
Encompass
Change view
Click to view available options
Quantity:
10 x 1 mL ampules
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88623 10 x 1 mL ampules
PI88625 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI88623 Supplier Thermo Scientific™ Supplier No. TS88623
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Employ the strongest silylator available for carbohydrates and steroids with the Thermo Scientific™ TMSI Silylation Reagent.

Employ the strongest silylator available for carbohydrates and steroids with the Thermo Scientific™ TMSI Silylation Reagent. This silylation reagent reacts quickly with hydroxyls and carboxylic acids but not with amines, and is particularly well suited in multiderivatization schemes for compounds containing both hydroxyl and amine groups. TMSI may be used to derivatize alcohols, phenols, organic acids, hormones, glycols, nucleotides, and narcotics and is excellent for C1–C5 fatty acids in serum and urine.

  • Reacts quickly and smoothly with hydroxyls and carboxylic acids but not with amines
  • Especially useful in multiderivatization schemes for compounds containing both hydroxyl and amine groups
  • Used in the derivatization of alcohols, phenols, organic acids, steroids, hormones, glycols, nucleotides and narcotics
  • Excellent for C1 through C5 fatty acids in serum and urine
TRUSTED_SUSTAINABILITY

Specifications

For Use With GC/MS
Standard Concentration 98 to 100%
Hazards Flammable, Irritant
Item Description 10 x 1 mL ampules
Storage 2°C to 8°C
Quantity 10 x 1 mL ampules
Standard Components 1-(Trimethylsilyl)-1 H-imidazole

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.