Learn More
Description
Employ the strongest silylator available for carbohydrates and steroids with the Thermo Scientific™ TMSI Silylation Reagent. This silylation reagent reacts quickly with hydroxyls and carboxylic acids but not with amines, and is particularly well suited in multiderivatization schemes for compounds containing both hydroxyl and amine groups. TMSI may be used to derivatize alcohols, phenols, organic acids, hormones, glycols, nucleotides, and narcotics and is excellent for C1–C5 fatty acids in serum and urine.
- Reacts quickly and smoothly with hydroxyls and carboxylic acids but not with amines
- Especially useful in multiderivatization schemes for compounds containing both hydroxyl and amine groups
- Used in the derivatization of alcohols, phenols, organic acids, steroids, hormones, glycols, nucleotides and narcotics
- Excellent for C1 through C5 fatty acids in serum and urine
Specifications
Specifications
| For Use With | GC/MS |
| Hazards | Flammable, Irritant |
| Storage | 2°C to 8°C |
| Quantity | 25 g |
| Standard Components | 1-(Trimethylsilyl)-1 H-imidazole |
| Standard Concentration | 98 to 100% |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.