Learn More
Description
Product # 90075 binds to immobilized TMT surface. Product # 90075 has been succesfully used in immunoprecipitation and Western blot applications.
Specifications
Specifications
| Antigen | TMT |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 25D5 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4% BSA and Kathon |
| Gene Alias | Tandem Mass Tag |
| Host Species | Mouse |
| Purification Method | Protein G |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
