Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMTC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555466 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555466 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555466 Supplier Invitrogen™ Supplier No. PA555466
  • $626.00
    $508.65 / Each of 1

    Save $117.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Immunogen sequence: MMGEAYMRLS KLPEAEHWYM ESLRSKTDHI PAHLTYGKLL ALTGRKSEAE KLFLKAIELD PTKGNCYMHY GQFLLEEARL IEAAEMAKKA AELDSTEFDV VFNAAHMLRQ ASLNEAAEKY YDLAARLRPN YPAALMNLGA IL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 94%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TMTC2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMTC2
Gene Accession No. Q56A06, Q8N394
Gene Alias 8430438D04Rik; D330034A10Rik; IBDBP1; IBDBP1 transcript 2; Protein O-mannosyl-transferase TMTC2; RGD1309848; TMTC2; transmembrane and tetratricopeptide repeat containing 2; transmembrane and TPR repeat-containing protein 2
Gene Symbols TMTC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMTC2. Recombinant protein control fragment (Product #RP-90774).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160335, 278279, 299762
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.