Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113838
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113838 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113838 Supplier Invitrogen™ Supplier No. PA5113838
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IRMGLLYITL CIVFLMTCKP PLYMGPEYIK YFNDKTIDEE LERDKRVTWI For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 85%.

This gene encodes a member of the disulfide isomerase (PDI) family of endoplasmic reticulum (ER) proteins that catalyze protein folding and thiol-disulfide interchange reactions. The encoded protein has an N-terminal ER-signal sequence, a catalytically active thioredoxin domain, one transmembrane domain and a C-terminal ER-retention sequence. This protein is enriched on the mitochondria-associated-membrane of the ER via palmitoylation of two of its cytosolically exposed cysteines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMX2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMX2
Gene Accession No. Q9Y320
Gene Alias 2310042M24Rik; AA589631; Cell proliferation-inducing gene 26 protein; CGI-31; growth-inhibiting gene 11; My009; PDIA12; PIG26; protein disulfide isomerase family A, member 12; PSEC0045; thioredoxin domain containing 14; thioredoxin domain-containing protein 14; thioredoxin related transmembrane protein 2; thioredoxin-related transmembrane protein 2; thioredoxin-related transmembrane protein-like protein; TMX2; TXNDC14; Unknown (protein for MGC:134152); UNQ237/PRO270
Gene Symbols TMX2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TXNDC14.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51075
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.