Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMX4 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571093
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1093 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-1093 Supplier Bethyl Laboratories, Inc Supplier No. A305724AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

TXNDC13 is a single-pass type I membrane protein. It contains 1 thioredoxin domain. The exact function of TXNDC13 remains unknown.

Specifications

Antigen TMX4
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene TMX4
Gene Accession No. Q9H1E5
Gene Alias DJ971N18.2, KIAA1162, PDIA14, protein disulfide isomerase family A, member 14, thioredoxin domain containing 13, thioredoxin domain-containing protein 13, TXNDC13
Gene Symbols TMX4
Host Species Rabbit
Immunogen Between 299 and 349
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56255
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.