Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tn Antigen Monoclonal Antibody (Tn 218)
GREENER_CHOICE

Catalog No. MA190544 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA190544 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA190544 Supplier Invitrogen™ Supplier No. MA190544
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA1-90544 stains Tn antigen in epithelial carcinoma, pancreatic, colon, stomach and breast. A suggested positive control for this product is human gastrointestinal tumor. The antibody is supplied as concentrated tissue culture supernatant in PBS with 0.05% azide.

Aberrant glycosylation is a hallmark of cancer cells. The blood group precursors T (Thomsen-Friedenreich) and Tn epitopes are shielded in healthy and benign-diseased tissues but uncovered in approximately 90% of carcinomas. T and Tn glycoproteins are specific, autoimmunogenic pancarcinoma antigens. These antigens may also be found in neoplastic blood cells. Fundamental, chemical and physical aspects of these glycoproteins of primary carcinomas are discussed first. Tn and T epitopes are cell and tissue adhesion molecules, essential in invasion by and metastasis of carcinoma which includes adherent and proliferative phases. Immunohistochemical studies of the extent of Tn antigen expression in primamry breast carcinoma demonstrate its highly significant correlation with clinical pathological tumor stages, and hence its value as a reliable prognisticator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tn Antigen
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone Tn 218
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.2
Gene Alias Thomsen-Friedenreich antigen
Host Species Mouse
Immunogen Asialo ovine submaxillary mucin (AOSM).
Purification Method Purified
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.