Learn More
Enzo Life Sciences TNF-alpha Recombinant monoclonal antibody (D2E7) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS5650200
Encompass_Preferred
Clone: D2E7 (Adalimumab). Isotype: IgGκ. Immunogen: This antibody was generated by affinity maturation from the 2SD4 scFv, which in turn was selected on human TNF alpha, using the murine anti-hTNF alpha antibody Mab32 as a template. Species reactivity: Human. Specificity: Recognizes soluble TNF-α. Applications: ELISA, IF, IHC, FUNC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3091
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.