Learn More
Diagnostic Biosystems TNF-ALPHA TNF-Alpha (Tumor Necrosis Factor alpha)
Supplier:  Diagnostic Biosystems MOB50201
Encompass_Preferred
This MAb recognizes human 17-26kDa protein, which is identified as cytokine TNF-α (Tumor Necrosis Factor-alpha). Monomeric human TNF-α is a 157 amino acid protein (non-glycosylated) with a reported molecular weight of 17 kDa and can be expressed as a free molecule, also TNF-α is generated as a precursor form called transmembrane TNF-α can be expressed as a cell surface type II polypeptide consisting of 233 amino acid residues molecular weight 26 kDa. TNF-α is an important cell-signaling component of the immune system. It is a protein secreted by LPS stimulated macrophages, and causes tumor necrosis when injected into tumor bearing mice. TNF-α is currently being evaluated in treatment of certain cancers and AIDS Related Complex.
Catalog No. 50-238-5567
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.