Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF-alpha Antibody, FITC, R&D Systems™
SDP

Catalog No. NBP119532F
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP119532F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP119532F Supplier R&D Systems Supplier No. NBP119532F

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

TNF-alpha Polyclonal specifically detects TNF-alpha in Canine, Fish, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen TNF-alpha
Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating
Formulation PBS with 0.05% Sodium Azide
Gene Alias APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Gene Symbols TNF
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human TNF alpha protein (between residues 100-200) [Uniprot: P01375]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 7124
Target Species Canine, Fish, Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.