Learn More
Genscript Corporation Antibody Protein Proteins Proteins 10 UG 1/EA
Supplier:  Genscript Corporation Z0331610
Encompass_Preferred
Tumor Necrosis Factor-Alpha (TNF-Alpha) plays a major role in regulating growth differentiation inflammation viral replication tumorigenesis and autoimmune diseases. TNF alpha-1a is a potent lymphoid factor that exerts cytotoxic effects on a wide range of tumor cells. In addition to inducing hemorrhagic necrosis of tumors studies indicate TNF is involved in tumor igenesis tumor metastasis viral replication septic shock fever inflammation Crohn s disease rheumatoid arthritis and graft-versus-host disease.
Catalog No. 50-252-9664
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.