Learn More
Enzo Life Sciences TNF-alpha (human), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ADI908066010
Encompass_Preferred
Alternative name: Tumor necrosis factor alpha, TNFSF 2. Purity: ≥98% (SDS-PAGE; RP-HPLC). Formulation: Lyophilized. Sterile filtered liquid, pH 7.2, containing 20mM PBS and 10mM sodium chloride. Source: Produced in E. coli. Long Term Storage: -20°C. Use/Stability: Upon reconstitution TNF-α should be stored at 4°C between 2-7 days and for future use below -20°C.  For long term storage it is recommended to add a carrier protein (0.1% HSA or BSA).  Please prevent freeze-thaw cycles. Applications: ELISA, WB in vitro Assay.
Catalog No. 50-200-7092
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.