Learn More
Invitrogen™ Human TNF alpha Matched Antibody Pair
Description
The Human TNF-alpha Matched Antibody Pair Kit comes with pre-matched antibody pairs, standards, and streptavidin-HRP to develop your own enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human TNF-alpha. Buffer reagents and microplates needed to complete the ELISA reaction are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Specifications
Specifications
| Antigen | TNF alpha |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Biotin |
| Gene Accession No. | P01375 |
| Gene Alias | Cachectin, Tumor necrosis factor alpha, DIF |
| Gene Symbols | TNF |
| Quantity | 10 x 96 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.