Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNF alpha Monoclonal Antibody (TN3-19.12), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5013701
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-137-01 50 μg
50-137-02 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-137-01 Supplier Invitrogen™ Supplier No. 14742381
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Armenian Hamster Monoclonal Antibody

Description: The TN3-19.12 monoclonal antibody reacts with mouse, rat, and rabbit tumor necrosis factor-alpha (TNF alpha), but not with human TNF alpha. The TN3-19.12 antibody is a neutralizing antibody. Mouse TNF alpha is a 17 kDa factor produced by macrophages, monocytes, neutrophils, CD4+ T cells and NK cells. A 26 kDa form of TNF alpha is expressed as a membrane-bound molecule. TNF alpha is cytolytic and plays an important role in immune regulation. Dimers and trimers of TNF alpha have been observed. Applications Reported: The TN3-19.12 antibody has been reported for use in intracellular staining for flow cytometric analysis, cytokine neutralization, ELISPOT, IVCCA, and ELISA. Fluorochrome conjugated TN3-19.12 is recommended for use in intracellular staining and flow cytometry. Functional Grade purified TN3-19.12, cat. 16-7423, is recommended for use in functional assays. For ELISPOT capture, the alternative clone, 1F3F3D4 (cat No. 16-7325), is recommended. Applications Tested: The TN3-19.12 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of mouse or rat TNF alpha in combination with the biotinylated anti-mouse/rat TNF alpha polyclonal antibody (13-7341) for detection and recombit mouse TNF alpha (14-8321) or rat TNF alpha as the standard. Product 14-8320 is a bioactive protein, but is not recommended for rat TNF alpha ELISA.] A suitable range of concentrations of this antibody for ELISA capture is 1-4 μg/mL.

TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF alpha
Applications ELISA, Flow Cytometry, Functional Assay, Neutralization
Classification Monoclonal
Clone TN3-19.12
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Tnf
Gene Accession No. P06804, P16599
Gene Alias APC1 protein; Cachectin; C-domain 1; C-domain 2; cTNF; DADB-70P7.1; DIF; ICD1; ICD2; Intracellular domain 1; Intracellular domain 2; N-terminal fragment; NTF; RATTNF; Tnf; TNF alpha; TNF superfamily; TNF α; TNF, macrophage-derived; TNF, monocyte-derived; TNFA; TNF-a; TNFalpha; TNF-alpha; Tnfsf1a; TNFSF2; TNFα; TNLG1F; Tumor necrosis factor; tumor necrosis factor (TNF superfamily, member 2); tumor necrosis factor alpha; tumor necrosis factor alpha (cachetin); tumor necrosis factor alpha precursor; tumor necrosis factor ligand 1F; tumor necrosis factor ligand superfamily member 2; Tumor necrosis factor, membrane form; Tumor necrosis factor, soluble form; tumor necrosis factor-alpha; tumor necrosis factor-alpha precursor; tumor-necrosis factor; tumour necrosis factor
Gene Symbols Tnf
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21926, 24835
Target Species Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.