Learn More
Arbor Assays The DetectX TNF-alpha Mouse ELISA Kit quantitatively measures TNF-alpha in mouse serum and plasma.
SDP
Supplier:  Arbor Assays K091H1
Encompass_Preferred
The DetectX TNF-alpha Mouse ELISA Kit from Arbor Assays is a Sandwich ELISA designed to quantitatively measure mouse TNF-alpha present in serum, plasma, and tissue culture media This 3.75h assay has a sensitivity of 11.0 pg/mL and a standard curve ranging from 15.7 - 3000 pg/mL. A recombinant mouse TNF-alpha standard is provided to generate a standard curve. Samples are added into a microtiter plate coated with a polyclonal antibody to capture TNF-alpha present in the sample. After a 2-hours and plate washing, a biotinylated TNF-alpha antibody is added. After a 60 minutes and plate washing, peroxidase-conjugated streptavidin is added. After a 30 minutes and plate washing, the substrate which reacts with the bound peroxidase-conjugated streptavidin is added. After a short incubation, the reaction is stopped and the intensity of the generated color is detected in a plate reader capable of measuring 450 nm wavelength and the concentration of the TNF-alpha in the sample is calculated.
Catalog No. NC2684356
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.