Learn More
ACROBiosystems Recombinant Protein;Human TNFR2 / CD120b / TNFRSF1B Protein;HEK293;50UG;TN2,TNFRSF1B,CD120b,TBPII,TNF-R-II,TNF-R75,TNFBR,TNFR1B,TNFR2,TNF R2,TNFR80,p75,p75TNFR,TNFR II,TNFRII,TNFR,TNFR
Supplier:  ACROBiosystems TN2H522750UG
Encompass_Preferred
Recombinant Protein;Human TNFR2 / CD120b / TNFRSF1B Protein;50UG;Human TNFR2, His Tag (TN2-H5227) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Asp 257 (Accession # AAA36755).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26 kDa. The protein migrates as 35-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human TNF-alpha (Cat. No. TNA-H4211) at 5 g/mL (100 l/well) can bind Human TNFR2, His Tag (Cat. No. TN2-H5227). The EC50 of human TNFR2 is 647 ng/mL (QC tested).;Immobilized ActiveMax Human TNF-beta (Cat. No. TNB-H5214) at 5 g/mL (100 l/well),can bind Human TNFR2, His Tag (Cat. No. TN2-H5227) with a linear range of 2-62 ng/mL (Routinely tested).
Catalog No. 50-165-7759
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.