Learn More
Invitrogen™ TNFRH3 Monoclonal Antibody (509027)

Description
In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Fas, rmTNF RI, and rmTNF RII is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | TNFRH3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 509027 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Tnfrsf26 |
| Gene Accession No. | P83626 |
| Gene Alias | TNF receptor homolog 3; TNFR; Tnfrh3; Tnfrsf24; Tnfrsf26; Tumor necrosis factor receptor; Tumor necrosis factor receptor superfamily member 26; tumor necrosis factor receptor superfamily, member 24; tumor necrosis factor receptor superfamily, member 26 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.