Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TNFRSF18-Cell lysates (GITR) ELISA Kit

Catalog No. 502741193
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-274-1193 5 x 96 Tests
EHTNFRSF18C 96 Tests
50-255-0911 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-274-1193 Supplier Invitrogen™ Supplier No. EHTNFRSF18CLX10
Add to Cart
Add to Cart
Request a Quote

ELISA

The Human TNFRSF18-Cell lysates (GITR) ELISA quantitates Hu GITR in human cell or tissue lysates. The assay will exclusively recognize both natural and recombinant Hu GITR. Principle of the method The Human GITR solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Tumor necrosis factor receptor superfamily member 18 (TNFRSF18), also called GITR or AITR is a protein that in humans is encoded by the TNFRSF18 gene. It is mapped to 1p36.33. This gene encodes a member of the TNF-receptor superfamily. The encoded receptor has been shown to have increased expression upon T-cell activation, and it is thought to play a key role in dominant immunological self-tolerance maintained by CD25 (+)CD4 (+) regulatory T cells. Knockout studies in mice also suggest the role of this receptor is in the regulation of CD3-driven T-cell activation and programmed cell death. Three alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y5U5
Assay Range 13.7 to 10,000 pg/mL
Assay Sensitivity 15 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates
Target Species Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 8784
Gene Symbol AITR,CD357,GITR,GITR-D
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 20 μL
Storage Requirements 2°C to 8°C
Target GITR
Test Time 1 hr. 20 min.
Total Assay Time 4 hr. 45 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.