Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TNFRSF6B (DcR3) ELISA Kit

Catalog No. 502741195
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-274-1195 10 x 96 Tests
EHTNFRSF6B 96 Tests
50-255-0915 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-274-1195 Supplier Invitrogen™ Supplier No. EHTNFRSF6BX10
Add to Cart
Add to Cart
Request a Quote

ELISA

The Human TNFRSF6B (DcR3) ELISA quantitates Hu DcR3 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu DcR3. Principle of the method The Human DcR3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95407
Assay Range 0.328-80 ng/mL
Assay Sensitivity 0.3 ng/mL
Calibration Range 0.3 ng/mL to 80 ng/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA antibody pair detects human DcR3. Other species not determined.
Sufficient For 10 x 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias DcR3; Decoy receptor 3; decoy receptor 3 variant 1; decoy receptor 3 variant 2; decoy receptor for Fas ligand; DJ583P15.1.1; hCG_2044836; M68; M68E; OTTHUMP00000031583; TNF receptor superfamily member 6b; TNFRSF6B; TR6; tumor necrosis factor receptor superfamily member 6B; tumor necrosis factor receptor superfamily member 6b, decoy; tumor necrosis factor receptor superfamily, member 6b, decoy; UNQ186/PRO212DcR3; Decoy receptor 3; decoy receptor 3 variant 1; decoy receptor 3 variant 2; decoy receptor for Fas ligand; DJ583P15.1.1; hCG_2044836; M68; M68E; OTTHUMP00000031583; TNF receptor superfamily member 6b; TNFRSF6B; TR6; tumor necrosis factor receptor superfamily member 6B; tumor necrosis factor receptor superfamily member 6b, decoy; tumor necrosis factor receptor superfamily, member 6b, decoy; UNQ186/PRO212; DCR3; DJ583P15.1.1; M68; M68E; TR6
Gene ID (Entrez) 8771
Gene Symbol TNFRSF6B
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases Decoy receptor 3, DcR3, Decoy receptor for Fas ligand, M68
Quantity 10 x 96 Tests
Regulatory Status RUO
Research Discipline Apoptosis
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym hCG
Target DcR3
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.