Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNNI3K Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106737
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106737 Supplier Invitrogen™ Supplier No. PA5106737
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TNNI3K.

TNNI3K may play a role in cardiac physiology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNNI3K
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TNNI3K
Gene Accession No. Q59H18, Q5GIG6, Q7TQP6
Gene Alias cardiac ankyrin repeat kinase; Cardiac troponin I-interacting kinase; Cark; CCDD; D830019J24Rik; Serine/threonine-protein kinase TNNI3K; si:dkey-267j14.4; TNNI3 interacting kinase; TNNI3-interacting kinase; Tnni3k; tnni3k {ECO:0000250; UniProtKB:Q59H18}
Gene Symbols TNNI3K
Host Species Rabbit
Immunogen A synthesized peptide derived from human TNNI3K(Accession Q59H18), corresponding to amino acid residues H205-L255.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 295531, 435766, 51086
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.