Learn More
Description
This antibody detects a band of approximately 90 kDa.
Specifications
Specifications
| Antigen | TNPO1 |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | D45 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | TNPO1 |
| Gene Accession No. | Q8BFY9, Q92973 |
| Gene Alias | AU021749; D13Ertd688e; importin 2; importin beta 2; importin beta-2; IPO2; karyopherin (importin) beta 2; karyopherin beta-2; KPNB2; M9 region interaction protein; MAPKAP1; MIP; MIP1; MSIN1; SAPK 1; SIN1; SIN-1; SIN1g; TNPO1; transportin; transportin 1; transportin-1; TRN; Unknown (protein for MGC:127455) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.