Learn More
Promega TNT™ T7 Quick Starter Bundle, Colorimetric
Catalog No. PRL1215 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
Catalog No. Quantity
PRL1215 1/Ea.
1 options
Catalog No. PRL1215 Supplier Promega Supplier No. L1215
Only null left
Add to Cart
Add to Cart
Includes
  • TNT™ T7 Quick Coupled Transcription/Translation System
  • Transcend™ Colorimetric Translation Detection System
  • pTNT™ Vector
  • pCMVTNT™ Vector
  • Transcend™ tRNA
Purchase this bundle, and get the TNT T7 Quick Coupled Transcription/Translation System, your choice of Transcend Translation Detection System, and receive two cell-free expression-qualified expression vectors, pTNT and pCMVTNT Vectors, at no extra cost.
  • Starter Pack for Protein Expression, Detection and Interactions Analysis
  • TnT(R) T7 Quick Coupled Transcription/Translation System, a simple one-hour, one-tube reaction
  • Transcend™ Translation Detection Systems detects proteins with either chemiluminescent or colorimetric techniques
  • pTnT™ and pCMVTnT™ Vectors work to enhance cell-free expression
Specifications
Content And Storage COMPONENTS AT DIFFERENT TEMPS
Quantity 1/Ea.