Learn More
Biotium TO iodide (515/531), 1 mM in DMSO
Supplier:  Biotium 40088
Encompass_Preferred
TO iodide, also know as TO-PRO-1, is a cell-impermeant, high-affinity carbocyanine monomeric nucleic acid stain that is essentially nonfluorescent in the absence of nucleic acids but exhibits green fluorescence when bound to nucleic acids. TO iodide is also used as a nuclear counterstain and dead cell indicator. Properties: Orange solution; Ex/Em= 515/531 nm (with DNA); Store at -20°C and protect from light; C24H29I2N3S, MW: 645.39.
Catalog No. 50-196-4666
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.