Learn More
Description
14915-1-AP recognizes 38 and 45 kDa bands. The additional 45 kDa band may be due to phosphorylation.
Specifications
Specifications
| Antigen | TOB1 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.29 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | TOB1 |
| Gene Accession No. | P50616, Q61471, Q8R5K6 |
| Gene Alias | APRO6, PIG49, Protein Tob1, TOB, TOB1, Transducer of erbB 2 1, transducer of ERBB2, 1, TROB, TROB1 |
| Gene Symbols | Tob1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
