Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tobramycin, MP Biomedicals™

Catalog No. ICN15692680 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
0.1 g
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15692680 0.1 g
ICN15692625 25 mg
ICN15692690 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15692680 Supplier MP Biomedicals, Inc Supplier No. 0215692680
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32986-56-4
Molecular Formula C18H37N5O9
Molecular Weight (g/mol) 467.52
InChI Key NLVFBUXFDBBNBW-PBSUHMDJSA-N
Synonym Distobram, Gernebcin
PubChem CID 36294
ChEBI CHEBI:28864
IUPAC Name (2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N

Specifications

Type Tobramycin
Physical Form Powder
Color White
Quantity 0.1 g
Formula Weight 467.5
Chemical Name or Material Tobramycin
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.