Learn More
Aqua Solutions TOC STANDARD 1000 PPM 100ML
Supplier: Aqua Solutions 9570100ML
Total Organic Carbon Standard; 1000mg/L (ppm) Solution Prepared to EPA 415 / Standard Methods; Clear colorless liquid; 100mL; Specific Gravity 1; Percent Volatile by Volume: > 99; Handle using safe laboratory practices