Learn More
TOC Standard, According to ISO/CEN EN1484, 500 mg/L±10 mg/L, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Concentration | 500 mg/L ±10 mg/L |
| Quantity | 500 mL |
| Grade | Analytical Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.