Learn More
Tocris Bioscience™ 2-Furoyl-LIGRLO-amide
GREENER_CHOICE
Catalog No. 30151
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
30-151 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 30-151 Supplier Tocris Bioscience™ Supplier No. 3015/1
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Potent and selective PAR2 agonist
2-Furoyl-LIGRLO-amide is a potent and selective PAR2 receptor agonist (pD2 = 7.0). Causes a dose-dependent relaxation of murine femoral arteries.
Specifications
Purity 0.95
Quantity 1 mg