Learn More
Tocris Bioscience™ ACTH (1-39)
GREENER_CHOICE
Catalog No. 34921
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
34-921 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 34-921 Supplier Tocris Bioscience™ Supplier No. 3492/1
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Potent endogenous MC2 agonist
ACTH (1-39) is a potent endogenous melanocortin receptor 2 (MC2) agonist (EC50 = 57 pM). Component of the hypothalamic-pituitary-adrenal (HPA) axis that stimulates glucocorticoid production and release from the adrenal cortex. Induces insulin resistance, promotes a proinflammatory profile and stimulates UCP-1 in adipocytes in vitro.
Specifications
Quantity 1 mg