Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ω-Agatoxin IVA
GREENER_CHOICE

Catalog No. 279910U
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-9910U 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 27-9910U Supplier Tocris Bioscience™ Supplier No. 2799/10U
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

CaV2.1 blocker

ω-Agatoxin IVA is a selective blocker of P-type calcium channels (IC50 < 1 - 3 nM). Also inhibits N-type channels at micromolar concentrations.

Specifications

Purity 95%
Content And Storage Store at -20°C
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.