Learn More
Tocris Bioscience™ Akt/SKG Substrate Peptide
Catalog No. 13531
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
13-531 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 13-531 Supplier Tocris Bioscience™ Supplier No. 1353/1
This item is not returnable. View return policy
Akt/PKB substrate (synthetic)
Akt/SKG Substrate Peptide is a synthetic peptide suitable as a substrate for Akt/PKB. Unlike other substrates, it is not phosphorylated by p70 S6 kinase or MAP kinase activated protein kinase-1.
Specifications
Quantity 1 mg