Learn More
Tocris Bioscience™ Amylin
GREENER_CHOICE
Catalog No. 3418500U
Change view
Click to view available options
Quantity:
500 μg
Catalog No. Quantity
34-185-00U 500 μg
1 options
This item is not returnable. View return policy
Catalog No. 34-185-00U Supplier Tocris Bioscience™ Supplier No. 3418/500U
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Endogenous peptide agonist for amylin, calcitonin, CGRP and adrenomedullin receptors
Amylin is an endogenous peptide agonist for amylin, calcitonin, CGRP and adrenomedullin receptors. Inhibits glucagon secretion, delays gastric emptying and acts as a satiety agent. Displays glucose lowering effects in vivo.
Specifications
Quantity 500 μg