Learn More
Tocris Bioscience™ Angiotensin (1-7)
Catalog No. 15621
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
15-621 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 15-621 Supplier Tocris Bioscience™ Supplier No. 1562/1
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Vasorelaxant peptide
Angiotensin (1-7) is an endogenous peptide fragment. Induces vasorelaxation through release of NO and prostaglandins, perhaps through activation of a non-AT1, non-AT2 receptor, Mas. Counteracts the vasoconstrictive and proliferative effects of angiotensin II and stimulates vasopressin (anti-diuretic hormone) release in vivo.
Specifications
Quantity 1 mg