Learn More
Tocris Bioscience™ Anti-PIKFyve
Catalog No. 4082100U
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
40-821-00U 100 μg
1 options
This item is not returnable. View return policy
Catalog No. 40-821-00U Supplier Tocris Bioscience™ Supplier No. 4082/100U
This item is not returnable. View return policy
Antibody recognizing PIKfyve
Antibody raised against recombinant protein corresponding to amino acid residues 1083-1200 of human PIKfyve (Accession No. Q9Y2I7). Sheep polyclonal; affinity purified. Recognizes Human and Rat PIKfyve. Not yet tested in other species.
Western Blot legend : (1:1000 dilution) staining of lysates from HEK293 cells transfected with HA-PIKfyve expressing constructs (lanes 1 and 3) and untransfected (lanes 2 and 4).