Learn More
Tocris Bioscience™ APC 366
GREENER_CHOICE
Catalog No. 25115
Change view
Click to view available options
Quantity:
5 mg
Catalog No. Quantity
25-115 5 mg
1 options
This item is not returnable. View return policy
Catalog No. 25-115 Supplier Tocris Bioscience™ Supplier No. 2511/5
Add to Cart
Add to Cart
This item is not returnable. View return policy
Tryptase inhibitor
APC 366 is a selective inhibitor of mast cell tryptase (Ki = 7.1 μM) that inhibits tryptase-induced histamine release from human tonsil and lung cells. Reduces airway inflammation and blocks postchallenge airway hyperresponsiveness in vivo.
Specifications
Purity 0.95
Quantity 5 mg